689378
Arkopal® N 100
Synonym(s):
4-Nonylphenyl-polyethylene glycol, Ethylene oxide-nonylphenol polymer, Polyethoxylated nonylphenol
About This Item
Recommended Products
form
liquid
mol wt
average mol wt 680
impurities
≤1.0% water
CMC
0.059 mM (20-25°C)
viscosity
350-450 mPa.s(20 °C)
transition temp
cloud point 45-50 °C
density
1.06 g/mL at 20 °C
InChI
1S/C19H32O3/c1-2-3-4-5-6-7-8-9-18-10-12-19(13-11-18)22-17-16-21-15-14-20/h10-13,20H,2-9,14-17H2,1H3
InChI key
BLXVTZPGEOGTGG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service