Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

688940

Sigma-Aldrich

1-Methyl-3-propylimidazolium methyl carbonate solution

~50% in methanol

Synonym(s):

PMIM MeOCO2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16N2O3
CAS Number:
Molecular Weight:
200.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

liquid

concentration

~50% in methanol

impurities

≤0.05% water

refractive index

n20/D 1.406

anion traces

bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤10 mg/kg
fluoride (F-): ≤10 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

SMILES string

COC([O-])=O.CCC[n+]1ccn(C)c1

InChI

1S/C7H13N2.C2H4O3/c1-3-4-9-6-5-8(2)7-9;1-5-2(3)4/h5-7H,3-4H2,1-2H3;1H3,(H,3,4)/q+1;/p-1

InChI key

ABSPGMJUOBVQMZ-UHFFFAOYSA-M

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

target_organs

Eyes,Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

78.8 °F

flash_point_c

26 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service