Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

688657

Sigma-Aldrich

Isobutyl acetoacetate

Arxada quality

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOCH2CH(CH3)3
CAS Number:
Molecular Weight:
158.19
Beilstein/REAXYS Number:
1761405
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

quality

Arxada quality

manufacturer/tradename

Arxada AG

impurities

≤0.10% water
≤0.20% acid (as acetic acid)
≤3.0% 2-butanol

refractive index

n20/D 1.424 (lit.)

bp

100 °C/22 mmHg (lit.)

density

0.98 g/mL at 25 °C (lit.)

functional group

ester
ketone

SMILES string

CC(C)COC(=O)CC(C)=O

InChI

1S/C8H14O3/c1-6(2)5-11-8(10)4-7(3)9/h6H,4-5H2,1-3H3

InChI key

ZYXNLVMBIHVDRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

172.4 °F - closed cup

flash_point_c

78 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service