Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

688444

Sigma-Aldrich

DL-2-Piperidine-d9 carboxylic acid

97 atom % D, 97% (CP)

Synonym(s):

DL-Pipecolic acid-(piperidine-d9)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D9H2NO2
Molecular Weight:
138.21
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

97 atom % D

assay

97% (CP)

mp

272 °C (lit.)

mass shift

M+9

SMILES string

[2H]C1([2H])NC([2H])(C(O)=O)C([2H])([2H])C([2H])([2H])C1([2H])[2H]

InChI

1S/C6H11NO2/c8-6(9)5-3-1-2-4-7-5/h5,7H,1-4H2,(H,8,9)/i1D2,2D2,3D2,4D2,5D

InChI key

HXEACLLIILLPRG-UHUJFCCWSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Hartmann et al.
Cell, 173(2), 456-469 (2018-03-27)
Following a previous microbial inoculation, plants can induce broad-spectrum immunity to pathogen infection, a phenomenon known as systemic acquired resistance (SAR). SAR establishment in Arabidopsis thaliana is regulated by the Lys catabolite pipecolic acid (Pip) and flavin-dependent-monooxygenase1 (FMO1). Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service