Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

688223

Sigma-Aldrich

trans-1,2-Dibromocyclopentane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8Br2
CAS Number:
Molecular Weight:
227.93
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.550

density

1.857 g/mL at 25 °C

SMILES string

Br[C@H]1CCC[C@@H]1Br

InChI

1S/C5H8Br2/c6-4-2-1-3-5(4)7/h4-5H,1-3H2/t4-,5-/m0/s1

InChI key

LJCGLSZQABMYGU-WHFBIAKZSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service