Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

687774

Sigma-Aldrich

DuPhos/BPE Ligands Kit I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352000

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

Kit Components Also Available Separately

Product No.
Description
SDS

  • 665207(−)-1,2-Bis[(2S,5S)-2,5-dimethylphospholano]ethane, kanata purity 100 mgSDS

  • 665266(+)-1,2-Bis[(2S,5S)-2,5-dimethylphospholano]benzene, kanata purity 100 mgSDS

  • 667854(+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane, kanata purity 100 mgSDS

  • (−)-1,2-Bis[(2S,5S)-2,5-diisopropylphospholano]benzene, kanata purity 100 mg

  • 6684351,2-Bis[(2S,5S)-2,5-diisopropylphospholano]ethane, kanata purity 100 mgSDS

  • 668451(−)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]ethane, kanata purity 100 mgSDS

  • 668486(+)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene, Namena purity 100 mgSDS

  • 1,1′-Bis[(2S,5S)-2,5-dimethylphospholano]ferrocene, kanata purity 100 mg

  • 677035(2S,5S)-(+)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane, kanata purity 100 mgSDS

  • 6785621,2-Bis[(2S,5S)-2,5-dimethylphospholano]benzene monooxide, kanata purity 100 mgSDS

  • 1,1′-Bis[(2S,5S)-2,5-diethylphospholano]ferrocene 100 mg

  • 1,1′-Bis[(2S,5S)-2,5-diisopropylphospholano]ferrocene 100 mg

See All (12)

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Pyr. Liq. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service