Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

687219

Sigma-Aldrich

1-Pentynylboronic acid pinacol ester

90%

Synonym(s):

2-(1-Pentynyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19BO2
CAS Number:
Molecular Weight:
194.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

90%

form

liquid

refractive index

n20/D 1.4479

bp

68-72 °C/0.09 mmHg

density

0.9057 g/mL at 25 °C

SMILES string

CCCC#CB1OC(C)(C)C(C)(C)O1

InChI

1S/C11H19BO2/c1-6-7-8-9-12-13-10(2,3)11(4,5)14-12/h6-7H2,1-5H3

InChI key

RXNSNUUTGWELNS-UHFFFAOYSA-N

Application

1-Pentynylboronic acid pinacol ester can be used as:
  • A substrate in the copper-catalyzed decyanative cross-coupling reactions with aryl selenocyanate to yield unsymmetrically substituted selanes.[1]
  • A reactant in the palladium-catalyzed 1,2-aminoacylation, 1,2-amino-vinylation and -alkynylation reactions with various oxime esters.[2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

154.9 °F - closed cup

flash_point_c

68.3 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-Silver Dual Catalyzed Decyanative C-Se Cross-Coupling
Mukherjee N, et al.
Advanced Synthesis & Catalysis, 359(2), 329-338 (2017)
An umpolung approach to alkene carboamination: palladium catalyzed 1, 2-amino-acylation,-carboxylation,-arylation,-vinylation, and-alkynylation
Faulkner A, et al.
Journal of the American Chemical Society, 137(22), 7224-7230 (2015)
Copper-Silver Dual Catalyzed Decyanative C-Se Cross-Coupling
Mukherjee N, et al.
advanced synthesis and catalysis, 359(2), 329-338 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service