Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

687170

Sigma-Aldrich

3-(Trimethylsilyl)-2-propynal

97%

Synonym(s):

(Trimethylsilyl)propiolaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10OSi
CAS Number:
Molecular Weight:
126.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.444

density

0.854 g/mL at 25 °C

functional group

aldehyde

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C#CC=O

InChI

1S/C6H10OSi/c1-8(2,3)6-4-5-7/h5H,1-3H3

InChI key

LJRWLSKYGWLYIM-UHFFFAOYSA-N

Application

3-(Trimethylsilyl)-2-propynal can be used as a reactant to synthesize:
  • Silicon-containing polyfunctional chiral tertiary phosphine oxides, which are employed as building blocks to prepare polydentate amphiphilic ligands for the design of novel metal complex catalysts.[1]
  • 4-trimethylsilyl-1H-1,2,3-triazole-5-carbaldehyde by reacting with trimethylsilyl azide.[2]
  • 4-trimethylsilylethynyl-4H-pyran-3,5-dicarbaldehyde via trimerization reaction in the presence of DABCO (1,4-diazabicyclo-[2.2.2]octane) as a catalyst.[3]

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

84.9 °F

flash_point_c

29.4 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection by 1 H NMR of malonaldehyde as the key intermediate in the trimerization of 3-trimethylsilylprop-2-ynal to 4-trimethylsilylethynyl-4 H-pyran-3, 5-dicarbaldehyde
Mareev AV, et al.
Russ. J. Org. Chem., 44(10), 1551-1553 (2008)
1, 3-dipolar cycloaddition of trimethylsilyl azide to propynals and dimerization of 1H-1, 2, 3-triazole-5-carbaldehydes to tricyclic bis-hemiaminals
Demina MM, et al.
Russ. J. Org. Chem., 40(12), 1804-1809 (2004)
Regioselective addition of secondary phosphine oxides to 3-(trialkylsilyl)-and 3-(trialkylgermyl)-2-propynals
Gusarova NK, et al.
Journal of Organometallic Chemistry, 659(1-2), 172-175 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service