Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

686867

Sigma-Aldrich

5-Isoquinolineboronic acid

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8BNO2
CAS Number:
Molecular Weight:
172.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

≥95.0%

form

solid

mp

166-188 °C

Storage temp.

2-8°C

SMILES string

OB(O)c1cccc2cnccc12

InChI

1S/C9H8BNO2/c12-10(13)9-3-1-2-7-6-11-5-4-8(7)9/h1-6,12-13H

Inchi Key

XKEYHBLSCGBBGU-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service