686476
Poly(styrene)-block-poly(ethylene glycol)
Synonym(s):
Polystyrene-polyethylene glycol
About This Item
Recommended Products
mol wt
Mn 21,000-30,000 (polystyrene)
Mn 21,000-31,000
Mn 700-1,100 (PEG)
Mw/Mn
1.1‑1.3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service