Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

686204

Sigma-Aldrich

Zirconium bromonorbornanelactone carboxylate triacrylate

Synonym(s):

PRM30

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(BrC8H8O2CO2)m(CH2CHCO2)nZr, m~1, n~3
UNSPSC Code:
12352103
NACRES:
NA.23

form

powder

mol wt

average mol wt ~600

contains

500 ppm methyl hydroquinone as inhibitor

Application

Please see Aldrich Technical Bulletin AL-238 for additional application and usage information.
Zirconium-containing multifunctional acrylate useful for producing cured, transparent films with high refractive indices (up to ~1.63).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service