Key Documents
686115
Sigma-Aldrich
Reference Kit for Hydrogen Storage
About This Item
UNSPSC Code:
26111700
NACRES:
NA.23
Application
Materials included in the kit are designed to be used with the Hy-Energy′s PCTPro-2000 System. They also can be used for demonstration purposes and as standards during the development of novel hydrogen storage and battery materials.
Kit Components Also Available Separately
Product No.
Description
SDS
  • 685933Lanthanum-nickel alloy, hydrogen-storage grade 10 gSDS
  • 685968Lanthanum-nickel-cobalt alloy, hydrogen-storage grade 10 gSDS
  • 685976Mischmetal-nickel alloy, hydrogen-storage grade 10 gSDS
  • Titanium-manganese alloy, hydrogen-storage grade 10 g
Choose from one of the most recent versions:
Certificates of Analysis (COA)