Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

686093

Sigma-Aldrich

Hydrogen Storage Materials for Research, Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700

Application

Kit containing products for hydrogen storage applications to assist you in designing research-scale experiments.

Features and Benefits

Hydrogen content, XRD plots and metal purity data are available for all components.

Other Notes

Products do not ship in a single package. The components will be delivered as separate units.

Kit Components Only

Product No.
Description

  • Magnesium hydride, hydrogen-storage grade 10 g

  • Calcium hydride, chunks, +4 mesh, reagent grade, 95% 10 g

Kit Components Also Available Separately

Product No.
Description
SDS

  • 686034Lithium aluminum hydride, hydrogen-storage grade 10 gSDS

  • 686050Lithium amide, hydrogen-storage grade 10 gSDS

  • 686026Lithium borohydride, hydrogen-storage grade, ≥90% 10 gSDS

  • 201049Lithium hydride, powder, −30 mesh, ≥95% 10 gSDS

  • 399558Lithium nitride, −60 mesh, ≥99.5% 10 gSDS

  • 685984Sodium aluminum hydride, hydrogen-storage grade 10 gSDS

  • 686042Sodium amide, hydrogen-storage grade 10 gSDS

  • 686018Sodium borohydride, hydrogen-storage grade, 98% 10 gSDS

  • 223441Sodium hydride, dry, 90% 10 gSDS

  • 287717Borane-ammonia complex, technical grade, 90% 10 gSDS

See All (10)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A - Water-react 1

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service