Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

686085

Sigma-Aldrich

Titanium(III) chloride

hydrogen-storage grade, ≥99.99%

Synonym(s):

Titanium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl3
CAS Number:
Molecular Weight:
154.23
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
assay:
≥99.99%
grade:
hydrogen-storage grade
form:
crystals

grade

hydrogen-storage grade

assay

≥99.99%

form

crystals

mp

440 °C (dec.) (lit.)

SMILES string

Cl[Ti](Cl)Cl

InChI

1S/3ClH.Ti/h3*1H;/q;;;+3/p-3

InChI key

YONPGGFAJWQGJC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used for the vapor-phase formation of intermetallic compounds with ultrafine particle size.

Analysis Note

XRD plots and metal purity data are available upon request.

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Pyr. Sol. 1 - Skin Corr. 1B

supp_hazards

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gnana S Siluvai et al.
Biochemistry, 48(51), 12133-12144 (2009-11-20)
Sco-like proteins contain copper bound by two cysteines and a histidine residue. Although their function is still incompletely understood, there is a clear involvement with the assembly of cytochrome oxidases that contain the Cu(A) center in subunit 2, possibly mediating
Investigation into the effectiveness of surface treatment on poly(methyl methacrylate) when exposed in the mouth.
M D Murray
The Journal of prosthetic dentistry, 59(3), 368-373 (1988-03-01)
Catalytic oxidation of hydrazo derivatives promoted by a TiCl3/HBr system.
Eyal Drug et al.
Journal of the American Chemical Society, 129(45), 13784-13785 (2007-10-26)
C Noubactep et al.
Journal of hazardous materials, 132(2-3), 202-212 (2005-11-08)
The effectiveness of elemental iron (Fe(0)) to remove uranium (U) from the aqueous phase has been demonstrated. While the mitigation effect is sure, discrepancies in the removal mechanism have been reported. The objective of this study was to investigate the
Yan Zhang et al.
Organic & biomolecular chemistry, 9(13), 4977-4982 (2011-05-21)
N-N bond cleavage in hydrazines is widely used in the preparation of amines and thus occupies a significant place in organic synthesis. In this paper, we report a new method for the reductive cleavage of N-N bonds in hydrazines by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service