Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

684333

Sigma-Aldrich

tert-Butyldimethylsilyl (R)-(–)-glycidyl ether

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20O2Si
CAS Number:
Molecular Weight:
188.34
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

optical activity

[α]22/D -7.0°

refractive index

n20/D 1.4310

bp

195-199 °C/760 mmHg

density

0.870 g/mL at 25 °C

functional group

ether

storage temp.

2-8°C

SMILES string

CC(C)(C)[Si](C)(C)OC[C@H]1CO1

InChI

1S/C9H20O2Si/c1-9(2,3)12(4,5)11-7-8-6-10-8/h8H,6-7H2,1-5H3/t8-/m1/s1

InChI key

YANSSVVGZPNSKD-MRVPVSSYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

165.0 °F - closed cup

flash_point_c

73.9 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service