684058
(1S, 2S)-1,2-Bis(2-chlorophenyl)ethylenediamine dihydrochloride
97%
Synonym(s):
(1S, 2S)-1,2-Bis(2-chlorophenyl)-1,2-ethanediamine dihydrochloride
About This Item
Recommended Products
assay
97%
form
powder
optical activity
[α]22/D +31.0°, c = 1 in H2O
mp
230-235 °C
SMILES string
Cl.Cl.N[C@H]([C@@H](N)c1ccccc1Cl)c2ccccc2Cl
InChI
1S/C14H14Cl2N2.2ClH/c15-11-7-3-1-5-9(11)13(17)14(18)10-6-2-4-8-12(10)16;;/h1-8,13-14H,17-18H2;2*1H/t13-,14-;;/m0../s1
InChI key
YXGVUHANCVCDJJ-AXEKQOJOSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The Chin group is interested in computational and experimental approaches to understanding stereoselective recognition and catalysis. Their studies in weak forces (H-bonding, electronic and steric effects) has led to a highly efficient method for making limitless varieties of chiral vicinal diamines from the 'mother diamine' that are useful for developing stereoselective organocatalysts or transition metal-based catalysts as well as for developing drugs (Acc Chem Res (2012) p1345). The 'mother diamine' is also useful for making binol, monophos and binap analogs. The Chin group is also interested in using reversible covalent bonds for stereoselective recognition and L to D conversion of natural and non-natural amino acids (EJOC (2012) p229).
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service