Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

683221

Sigma-Aldrich

[Ru(benzene)Cl2]2

Umicore

Synonym(s):

Benzeneruthenium(II) chloride dimer, Dichloro(benzene)ruthenium(II)-dimer, Bis[(benzene)dichlororuthenium], Dibenzenetetrachlorodiruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ru2(C6H6)2Cl4
CAS Number:
Molecular Weight:
500.18
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

SMILES string

Cl[Ru]Cl.Cl[Ru]Cl.c1ccccc1.c2ccccc2

InChI

1S/2C6H6.4ClH.2Ru/c2*1-2-4-6-5-3-1;;;;;;/h2*1-6H;4*1H;;/q;;;;;;2*+2/p-4

InChI key

YGXMUPKIEHNBNQ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Hydrosilylation Catalysts

  • Catalyst for SNAr reactions of fluoroarenes and cyclic amines (eq. 1)
  • Precatalyst for the hydrogenation of β-keto esters (eq. 2)
  • Precursor for the preparation of other ruthenium catalysts (eq. 3)

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service