Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

683051

Sigma-Aldrich

2,6-Diisobutylaniline

96%

Synonym(s):

2,4-Bis(2-methylpropyl)benzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23N
CAS Number:
Molecular Weight:
205.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

liquid

refractive index

n20/D 1.513

density

0.898 g/mL at 25 °C

SMILES string

CC(C)Cc1cccc(CC(C)C)c1N

InChI

1S/C14H23N/c1-10(2)8-12-6-5-7-13(14(12)15)9-11(3)4/h5-7,10-11H,8-9,15H2,1-4H3

InChI key

PSIJIENWURFNAS-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

225.0 °F

flash_point_c

107.2 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service