682993
(11bR, 11′bR)-4,4′-(Oxydi-2,1-phenylene)bis-dinaphtho[2,1-d:, 1′, 2′-f][1,3,2]dioxaphosphepin
Synonym(s):
R, R-Reetz D-Diphosphonite
About This Item
Recommended Products
form
solid
optical activity
[α]22/D +123°, c = 1 in chloroform
mp
171-187 °C
SMILES string
O1P(Oc2ccc3ccccc3c2-c4c1ccc5ccccc45)c6ccccc6Oc7ccccc7P8Oc9ccc%10ccccc%10c9-c%11c(O8)ccc%12ccccc%11%12
InChI
1S/C52H32O5P2/c1-5-17-37-33(13-1)25-29-43-49(37)50-38-18-6-2-14-34(38)26-30-44(50)55-58(54-43)47-23-11-9-21-41(47)53-42-22-10-12-24-48(42)59-56-45-31-27-35-15-3-7-19-39(35)51(45)52-40-20-8-4-16-36(40)28-32-46(52)57-59/h1-32H
InChI key
UYGCVTAJLJBNHG-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Flam. Sol. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2
target_organs
Central nervous system
Storage Class
4.1B - Flammable solid hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service