All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C13H10O
CAS Number:
Molecular Weight:
182.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
assay
96%
form
solid
mp
45-50 °C
functional group
hydroxyl
SMILES string
OCC#Cc1cccc2ccccc12
InChI
1S/C13H10O/c14-10-4-8-12-7-3-6-11-5-1-2-9-13(11)12/h1-3,5-7,9,14H,10H2
Inchi Key
FARRJNMECLDKPD-UHFFFAOYSA-N
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
>230.0 °F - closed cup
flash_point_c
> 110 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service