Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

682748

Sigma-Aldrich

3-Nitrophenylboronic acid D-tartaric acid ester solution

1.0 M in THF

Synonym(s):

2-(3-Nitrophenyl)-1,3,2-dioxaborolane-4S,5S-dicarboxylic acid, D-TarB-NO2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8BNO8
Molecular Weight:
280.98
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

liquid

optical activity

[α]22/D -9°, neat

concentration

1.0 M in THF

refractive index

n20/D 1.4501

density

1.0351 g/mL at 25 °C

SMILES string

OC(=O)[C@H]1OB(O[C@@H]1C(O)=O)c2cccc(c2)[N+]([O-])=O

InChI

1S/C10H8BNO8/c13-9(14)7-8(10(15)16)20-11(19-7)5-2-1-3-6(4-5)12(17)18/h1-4,7-8H,(H,13,14)(H,15,16)/t7-,8-/m0/s1

InChI key

DKYPDMHYDHHGRL-YUMQZZPRSA-N

Application

In conjunction with NaBH4, this Lewis acid reduces prochiral alkyl aryl ketones with excellent enantioselectivity.[1][2]

signalword

Danger

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-0.0 °F

flash_point_c

-17.78 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kim, J. et al.
Organic Process Research & Development, 10, 949-949 (2006)
Kim, J.; Singaram, B.
Tetrahedron Letters, 47, 3901-3901 (2006)

Related Content

New asymmetric methodology meets the continuous demand for optically active compounds in various fields.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service