Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

682497

Sigma-Aldrich

1H-Indole-5-carboxamide

97%

Synonym(s):

5-(Aminocarbonyl)-1H-indole, 5-Carbamoylindole, 5-Carboxamidoindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2O
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

159-163 °C

SMILES string

NC(=O)c1ccc2[nH]ccc2c1

InChI

1S/C9H8N2O/c10-9(12)7-1-2-8-6(5-7)3-4-11-8/h1-5,11H,(H2,10,12)

InChI key

GQMYQEAXTITUAE-UHFFFAOYSA-N

application

  • Used for preparation of potential fructose bisphosphatase inhibitors
  • Reactant for preparation of (indolylamino)thienopyridine-carbonitriles using substitution and Suzuki cross-coupling as potential protein kinase Cθ inhibitors
  • Reactant for preparation of 5-substituted 3-(1′,2′,5′,6′-tetrahydropyridin-3′-yl) indoles as serotonin receptor binding agents

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service