Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

682306

Sigma-Aldrich

(R)-(–)-4,12-Bis[di(3,5-xylyl)phosphino]-[2.2]-paracyclophane

95%

Synonym(s):

(R)-5,11-Bis(3,5-xylylphosphino)tricyclo[8.2.2.24,7]hexadeca-hexaene, (R)-Xylyl-PHANEPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H50P2
CAS Number:
Molecular Weight:
688.86
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

optical activity

[α]22/D -61.0, c = 0.1 in ethanol

mp

234-238 °C

functional group

phosphine

SMILES string

Cc1cc(C)cc(c1)P(c2cc(C)cc(C)c2)c3cc4CCc5ccc(CCc3cc4)cc5P(c6cc(C)cc(C)c6)c7cc(C)cc(C)c7

InChI

1S/C48H50P2/c1-31-17-32(2)22-43(21-31)49(44-23-33(3)18-34(4)24-44)47-29-39-9-13-41(47)15-11-40-10-14-42(16-12-39)48(30-40)50(45-25-35(5)19-36(6)26-45)46-27-37(7)20-38(8)28-46/h9-10,13-14,17-30H,11-12,15-16H2,1-8H3

InChI key

LYHOBZAADXPPNW-UHFFFAOYSA-N

Application

Efficient ligand for asymmetric hydrogenation of dehydroamino acids, methyl esters and ketones.[1][2]

Legal Information

Sold in collaboration with Johnson Matthey Catalyst for research purposes only. US5874629 and any patents arising therefrom apply.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burk et al.
Organic letters, 2(26), 4173-4176 (2001-01-11)
PhanePhos-ruthenium-diamine complexes catalyze the asymmetric hydrogenation of a wide range of aromatic, heteroaromatic, and alpha, beta-unsaturated ketones with high activity and excellent enantioselectivity.
Journal of the American Chemical Society, 119, 6207-6207 (1997)

Articles

P-Phos ligand family enhances catalysis, featuring atropisomeric biaryl bisphosphine with unique structural elements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service