Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68215

Sigma-Aldrich

Dimethoxy(methyl)octylsilane

≥95.0% (GC)

Synonym(s):

Methyl-octyldimethoxysilane, Octylmethyldimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H26O2Si
CAS Number:
Molecular Weight:
218.41
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

vapor pressure

1.23 mmHg ( 54.4 °C)

assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.421 (lit.)

density

0.863 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC[Si](C)(OC)OC

InChI

1S/C11H26O2Si/c1-5-6-7-8-9-10-11-14(4,12-2)13-3/h5-11H2,1-4H3

InChI key

GOIPELYWYGMEFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Silica's versatility spans various industries, including biomedical applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service