Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

682047

Sigma-Aldrich

2-(Dimethylvinylsilyl)pyridine

90%

Synonym(s):

Dimethyl(2-pyridyl)vinylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NSi
CAS Number:
Molecular Weight:
163.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

90%

refractive index

n20/D 1.504

density

0.916 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C[Si](C)(C=C)c1ccccn1

InChI

1S/C9H13NSi/c1-4-11(2,3)9-7-5-6-8-10-9/h4-8H,1H2,2-3H3

InChI key

UAMVROIJEVLNFM-UHFFFAOYSA-N

Application

Participates in a variety of C-C and C-X bond-forming processes including carbomagnesiation and borylation.[1]

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

129.9 °F - closed cup

flash_point_c

54.4 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Itami, K.; Yoshida, J.
Synlett, 157-157 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service