Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

681954

Sigma-Aldrich

Methyl 3-methylpyridine-2-carboxylate

97%

Synonym(s):

3-Methylpicolinic acid, methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.516

density

1.121 g/mL at 25 °C

SMILES string

COC(=O)c1ncccc1C

InChI

1S/C8H9NO2/c1-6-4-3-5-9-7(6)8(10)11-2/h3-5H,1-2H3

InChI key

CCQFKEITVOTHIW-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service