Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

681512

Detergent 8®, liquid detergent

biodegradable

Synonym(s):

biodegradable detergent, non-ionic detergent, ultrasonic cleaner detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800

manufacturer/tradename

Alconox, Inc.

General description

Biodegradable; contains no phosphates. Free rinsing; leaves no residues. 1 gallon of concentrate makes up to 50 gallons of cleaning solution.

Legal Information

Detergent 8 is a registered trademark of Alconox, Inc.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service