Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

681504

Sigma-Aldrich

Pt (0) EnCat® 40

matrix encapsulaed nanoparticles, support (wet), extent of labeling: 0.12 mmol/g loading (dry basis)

Synonym(s):

Platinum (0), microencapsulated in polyurea matrix

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pt
UNSPSC Code:
12161600
NACRES:
NA.22

reaction suitability

core: platinum
reagent type: catalyst

extent of labeling

0.12 mmol/g loading (dry basis)

matrix

encapsulaed nanoparticles, support (wet)

Application

A safer and more user friendly alternative to classical Pt/C used in hydrogenation reactions; with minimal metal contamination by the catalyst which can be removed by filtration. Highly selective in the hydrogenation of nitro aromatics bearing halogen substitution.

Legal Information

EnCat is a registered trademark of Reaxa Ltd.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service