Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

681032

Sigma-Aldrich

3′-Chloro-4′-hydroxyacetophenone

95%

Synonym(s):

1-(3-Chloro-4-hydroxyphenyl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO2
CAS Number:
Molecular Weight:
170.59
Beilstein/REAXYS Number:
2043261
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

92-105 °C

functional group

chloro
ketone

SMILES string

CC(=O)c1ccc(O)c(Cl)c1

InChI

1S/C8H7ClO2/c1-5(10)6-2-3-8(11)7(9)4-6/h2-4,11H,1H3

InChI key

GMTSPBYBJKGPJF-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service