Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

680427

Sigma-Aldrich

QuadraSil® Metal Scavengers Kit I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.22

Legal Information

QuadraSil is a registered trademark of Johnson Matthey Finland Oy

Kit Components Also Available Separately

Product No.
Description
SDS

  • 679496QuadraSil® TA, extent of labeling: 0.5-1.5 mmol/g loading 5 gSDS

  • 679518QuadraSil® MTU, extent of labeling: 1.0-1.5 mmol/g loading 5 gSDS

  • 679526QuadraSil® MP, extent of labeling: 1.0-1.5 mmol/g loading 5 gSDS

  • 679534QuadraSil® AP, extent of labeling: 1.5-2.0 mmol/g loading 5 gSDS

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service