Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

680338

Sigma-Aldrich

2-(2-Hydroxyethylamino)-3-methylbutyric acid

97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO3
Molecular Weight:
163.21
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

assay

97% (CP)

SMILES string

CC(C)C(NCCO)C(O)=O

InChI

1S/C7H15NO3/c1-5(2)6(7(10)11)8-3-4-9/h5-6,8-9H,3-4H2,1-2H3,(H,10,11)

InChI key

AJNYQRXDVGKEIT-UHFFFAOYSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service