Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

679712

Sigma-Aldrich

S-Methyl N-(2,2,2-trichloroethoxysulfonyl)carbonchloroimidothioate

97%

Synonym(s):

N-(2,2,2-Trichloroethoxysulfonyl)carbonchlorothioic acid S methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5Cl4NO3S2
CAS Number:
Molecular Weight:
321.03
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

75-79 °C

functional group

chloro
thioether

SMILES string

CS\C(Cl)=N\S(=O)(=O)OCC(Cl)(Cl)Cl

InChI

1S/C4H5Cl4NO3S2/c1-13-3(5)9-14(10,11)12-2-4(6,7)8/h2H2,1H3/b9-3+

InChI key

NGCLYHLLOXVGSU-YCRREMRBSA-N

Application

Reactant for preparation of:
  • Muramycin analogs as antibacterial agents against drug-resistant bacteria
  • L-epicapreomycidine using rhodium-catalyzed stereoselective oxidative cyclization
  • (+)-gonyautoxin 3 via Rh-catalyzed amination
  • (+)-monobromophakellin and (+)-phakellin via a strategy applicable to synthesis of more complex members of this family of marine sponge-derived alkaloids including palau′amine
  • 2-imidazolidinones and 2-imino-1,3-imidazolidines
Reagent for preparation of protected guanidine derivatives.[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mihyong Kim et al.
Organic letters, 8(6), 1073-1076 (2006-03-10)
[reaction: see text] Oxidative C-H amination of N-trichloroethoxysulfonyl-protected ureas and guanidines is demonstrated to proceed in high yield for tertiary and benzylic-derived substrates. The success of these reactions is predicated on the choice of the electron-withdrawn 2,2,2-trichloroethoxysulfonyl (Tces) protecting group

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service