Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

679518

Sigma-Aldrich

QuadraSil® MTU

extent of labeling: 1.0-1.5 mmol/g loading

Synonym(s):

QuadraSil® Methylthiourea

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

extent of labeling

1.0-1.5 mmol/g loading

Quality Level

Application

Metal scavenger: Co, Cu, Fe, Pb, Pd, Rh, Ru, Ag, Au, Pt, Hg

Legal Information

QuadraSil is a registered trademark of Johnson Matthey Finland Oy

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service