Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

679313

Sigma-Aldrich

Trimethoxyphenylsilane

deposition grade, 98%

Synonym(s):

(Trimethoxysilyl)benzene, Phenyltrimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5Si(OCH3)3
CAS Number:
Molecular Weight:
198.29
Beilstein/REAXYS Number:
2937896
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

assay

98%

form

liquid

refractive index

n20/D 1.468 (lit.)

bp

233 °C (lit.)

density

1.062 g/mL at 25 °C (lit.)

SMILES string

CO[Si](OC)(OC)c1ccccc1

InChI

1S/C9H14O3Si/c1-10-13(11-2,12-3)9-7-5-4-6-8-9/h4-8H,1-3H3

InChI key

ZNOCGWVLWPVKAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - STOT RE 2 Oral

target_organs

Bladder

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

84.2 °F - closed cup

flash_point_c

29 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhe Dong et al.
Molecules (Basel, Switzerland), 24(3) (2019-01-27)
In this study, Candida rugosa lipase (CRL) was immobilized into modified hollow mesoporous silica (HMSS) materials with different hydrophobicity. Among propyl-(C₃), phenyl-(C₆), octyl-(C₈), and octadecyl-(C18) modified HMSS as well as native HMSS, taking advantage of more hydrophobic microenvironment, the HMSS-C18-CRL
Aleksandr A Goncharenko et al.
Molecules (Basel, Switzerland), 25(17) (2020-08-23)
The design of cargo carriers with high biocompatibility, unique morphological characteristics, and capability of strong bonding of fluorescent dye is highly important for the development of a platform for smart imaging and diagnostics. In this paper, BODIPY-doped silica nanoparticles were
Xiuhuan Song et al.
Polymers, 11(2) (2019-04-10)
A series of hybrid thermoplastic polyurethanes (PUs) were synthesized from bi-functional polyhedral oligomeric silsesquioxane (B-POSS) and polycaprolactone (PCL) using 1,6-hexamethylene diisocyanate (HDI) as a coupling agent for the first time. The newly synthesized hybrid materials were fully characterized in terms
Seiji Tsunega et al.
ChemPlusChem, 85(4), 619-626 (2020-04-03)
Systems that show circularly polarized luminescence (CPL) are usually constructed in one of two possible ways: either covalently binding the chiral moieties (usually organic compounds) to luminophores (inorganic or organic compounds) or associating the luminophores as guests with chiral hosts
Mohammad T Jafari et al.
Analytica chimica acta, 853, 460-468 (2014-12-04)
The capability of electrospray ionization-conventional ion mobility spectrometry (ESI-IMS) for direct analysis of the samples extracted by solid phase microextraction (SPME) was investigated and evaluated for the first time. To that end, an appropriate new desorption chamber was designed and

Articles

Deposition Grade Silanes, fully characterized by chemical analysis and nuclear magnetic resonance (NMR) with greater than 98% purity, for Sol-Gel Processes.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Synthesis of Melting Gels Using Mono-Substituted and Di-Substituted Alkoxysiloxanes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service