Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

679283

Sigma-Aldrich

PEDOT-Iron(III) oxide preparation

Synonym(s):

Poly(3,4-ethylenedioxythiophene) nanotubes-Iron(III) oxide composite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H4O2S)n : Fe2O3
UNSPSC Code:
12352103

form

solid

General description

Prepared following procedure described in Zhang, X., et al., Macromolecules 2006, 39, 470. The preparation includes nm-scale tubular PEDOT structures decorated with Iron(III) oxide.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service