Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

678589

Sigma-Aldrich

3-Methylfuran-2-carbonyl chloride

97%

Synonym(s):

3-Methyl-2-furoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5ClO2
CAS Number:
Molecular Weight:
144.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.5325

bp

70-70.5 °C/14 mmHg

density

1.263 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Cc1ccoc1C(Cl)=O

InChI

1S/C6H5ClO2/c1-4-2-3-9-5(4)6(7)8/h2-3H,1H3

InChI key

DFXNZMRXFMWEGQ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

190.0 °F - closed cup

flash_point_c

87.78 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service