Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

677736

Sigma-Aldrich

2-Bromo-3,5-dimethoxytoluene

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$77.90

$77.90

List Price$82.00
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 G
$77.90

About This Item

Empirical Formula (Hill Notation):
C9H11BrO2
CAS Number:
Molecular Weight:
231.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$77.90

List Price$82.00
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

95%

form

solid

mp

50-54 °C

functional group

bromo

SMILES string

COc1cc(C)c(Br)c(OC)c1

InChI

1S/C9H11BrO2/c1-6-4-7(11-2)5-8(12-3)9(6)10/h4-5H,1-3H3

InChI key

RONRLLIJBQHXCI-UHFFFAOYSA-N

Related Categories

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service