Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

677558

Sigma-Aldrich

(S)-(−)-2-[(1-Hydroxy-3,3-dimethylbutan-2-ylimino)methyl]-4,6-diiodophenol

97%

Synonym(s):

2-[[[(1S)-1-(Hydroxymethyl)-2,2-dimethylpropyl]imino]methyl]-4,6-diiodophenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17I2NO2
CAS Number:
Molecular Weight:
473.09
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

optical activity

[α]20/D -19°, c = 3% in acetone

mp

164-168 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)[C@@H](CO)\N=C\c1cc(I)cc(I)c1O

InChI

1S/C13H17I2NO2/c1-13(2,3)11(7-17)16-6-8-4-9(14)5-10(15)12(8)18/h4-6,11,17-18H,7H2,1-3H3/b16-6+/t11-/m1/s1

InChI key

WVBNUUNXQNMMOB-OHUUDMDPSA-N

Application

Useful in catalytic asymmetric sulfoxidation of alkyl aryl sulfides[1] and in the kinetic resolution of alkyl aryl sulfoxides.[2]

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanadium-catalyzed sulfur oxidation/kinetic resolution in the synthesis of enantiomerically pure alkyl aryl sulfoxides.
Carmelo Drago et al.
Angewandte Chemie (International ed. in English), 44(44), 7221-7223 (2005-10-18)
Pelotier, B. et al.
Synlett, 1055-1055 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service