Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

677442

Sigma-Aldrich

Samarium strontium cobalt oxide

nanopowder, <50 nm particle size (BET), 99.5% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
PubChem Substance ID:

description

SmSrCo4Ox

assay

99.5% trace metals basis

form

nanopowder

surface area

>25 m2/g

particle size

<50 nm (BET)

SMILES string

O=[Co].O=[Co].O=[Sr].O=[Co]O[Co]=O.O=[Co]O[Co]=O.O=[Sm]O[Sm]=O

InChI

1S/6Co.12O.2Sm.Sr

InChI key

QNWNHKBYDISCHS-UHFFFAOYSA-N

Application

Used as material component in high performance solid oxide fuel cells.[1]

Legal Information

Product of Engi-Mat Co.

pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction Model of Dense Sm0.5Sr0.5CoO3 as SOFC Cathode
H. Fukunaga, et al.
Solid State Ionics, 132, 279-285 (2000)

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service