Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

677434

Sigma-Aldrich

Silver-tin alloy

nanopowder, <150 nm particle size, 3.5% Ag basis, ≥97%

Synonym(s):

AgSn25

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgSn25
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

assay

≥97%
3.5% Ag basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: tin

surface area

>5.4 m2/g

particle size

<150 nm

SMILES string

[Ag].[Sn]

InChI

1S/Ag.Sn

Inchi Key

QCEUXSAXTBNJGO-UHFFFAOYSA-N

Pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Recent Advances of Conductive Adhesives as a Lead-Free Alternative in Electronic Packaging: Material, Processing, Reliability and Applications.
Y. Li, C.P. Wong
Materials Science & Engineering. R, Reports : A Review Journal, 51, 1-35 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service