Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

677361

Sigma-Aldrich

Polyepoxysuccinic acid

Synonym(s):

Polyoxirane-2,3-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H4O5)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Product 677361 is not currently sold in your country. Contact Technical Service

form

solid

solubility

H2O: soluble

Application

Scale inhibitor

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–5 of 5 Questions  
  1. Where is Product 677361, Polyepoxysuccinic acid, primarily used?

    1 answer
    1. Polyepoxysuccinic acid is widely used in water treatment plants.

      Helpful?

  2. What are the biodegradation properties of Product 677361, Polyepoxysuccinic acid?

    1 answer
    1. Polyepoxysuccinic acid has good biodegradation properties, it can be widely used in circulating cool water system in situation of high alkaline, high hardness and high pH value.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. What is the molecular weight of product number 677361, Polyepoxysuccinic acid?

    1 answer
    1. The supplier does not measure the molecular weight of this material and does not have an estimation either.

      Helpful?

  5. What is the main application for Product 677361, Polyepoxysuccinic acid?

    1 answer
    1. Polyepoxysuccinic Acid is a multivariate scale and corrosion inhibitor with non-phosphor and non-nitrogen.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service