Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

677108

Sigma-Aldrich

1,4-Cyclohexane dimethanol-d4

mixture of cis and trans isomers, 98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D4H12O2
Molecular Weight:
148.24
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

98 atom % D

assay

98% (CP)

bp

283 °C (lit.)

mass shift

M+4

SMILES string

[2H]C([2H])(O)C1CCC(CC1)C([2H])([2H])O

InChI

1S/C8H16O2/c9-5-7-1-2-8(6-10)4-3-7/h7-10H,1-6H2/i5D2,6D2

InChI key

YIMQCDZDWXUDCA-NZLXMSDQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

61.0 °F - closed cup

flash_point_c

16.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service