Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

677051

Sigma-Aldrich

(2R,5R)-(+)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane

kanata purity

Synonym(s):

R,R-Et-RajPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25O2P
Molecular Weight:
292.35
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

optical activity

[α]/D +35°, c = 1 in chloroform

refractive index

n20/D 1.562

density

1.073 g/mL

SMILES string

CC[C@@H]1CC[C@@H](CC)P1c2ccccc2C3OCCO3

InChI

1S/C17H25O2P/c1-3-13-9-10-14(4-2)20(13)16-8-6-5-7-15(16)17-18-11-12-19-17/h5-8,13-14,17H,3-4,9-12H2,1-2H3/t13-,14-/m1/s1

Inchi Key

UKBFKBMQNUOKTD-ZIAGYGMSSA-N

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.
RajPhos is a trademark of Kanata Chemical Technologies, Inc.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service