676667
B-Methoxy-10-trimethylsilyl-9-borabicyclo(3.3.2)decane
95%
Synonym(s):
10-TMS-9-BBD, 10-TMS-9-Borabicyclo[3.3.2]decane, 9-Methoxy-10-trimethylsilanyl-9-borabicyclo(3.3.2)decane
About This Item
Recommended Products
assay
95%
reaction suitability
reaction type: C-C Bond Formation
refractive index
n20/D 1.494
bp
85 °C/0.4 mmHg
density
0.934 g/mL at 25 °C
SMILES string
COB1C2CCCC(CCC2)C1[Si](C)(C)C
InChI
1S/C13H27BOSi/c1-15-14-12-9-5-7-11(8-6-10-12)13(14)16(2,3)4/h11-13H,5-10H2,1-4H3
Inchi Key
AUJACFLHZMIAEG-UHFFFAOYSA-N
Application
Reactant for:
- Palladium-catalyzed allylation reaction
- Hydroboration of allenes
- Stereoselective γ-methoxyallylation reactions
- Asymmetric carboborylation reactions
- Insertion and borotropic rearrangement reactions
- Asymmetric Michael addition reactions
- Asymmetric propargylation of aldehydes
- Asymmetric allenylboration reactions
signalword
Danger
hcodes
pcodes
Hazard Classifications
Flam. Liq. 3 - Water-react 2
supp_hazards
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk_germany
WGK 3
flash_point_f
138.9 °F - closed cup
flash_point_c
59.4 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service