Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

676586

Sigma-Aldrich

cis-1,2-Dichloroethylene-13C2

99 atom % 13C, 97% (CP)

Synonym(s):

cis-Acetylene-13C2 dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl13CH13CHCl
CAS Number:
Molecular Weight:
98.93
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

assay

97% (CP)

bp

60 °C (lit.)

mp

-80 °C (lit.)

density

1.310 g/mL at 25 °C (lit.)

mass shift

M+2

SMILES string

Cl\[13CH]=[13CH]/Cl

InChI

1S/C2H2Cl2/c3-1-2-4/h1-2H/b2-1-/i1+1,2+1

InChI key

KFUSEUYYWQURPO-GQTGYUFJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

43.0 °F - closed cup

flash_point_c

6.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service