Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

676551

Sigma-Aldrich

Potassium 2-(3,5-di-tert-butyl-2-hydroxybenzylideneamino)-2,2-diphenylacetate

95%

Synonym(s):

Potassium N-(3,5-di-tert-butylsalicylidene)-2-amino-2,2-diphenylacetate, SALDIPAC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H32KNO3
Molecular Weight:
481.67
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

reaction suitability

reagent type: ligand

mp

122.2-131.9 °C

SMILES string

CC(C)(C)c1cc(\C=N\C(C(=O)O[K])(c2ccccc2)c3ccccc3)c(O)c(c1)C(C)(C)C

InChI

1S/C29H33NO3.K/c1-27(2,3)23-17-20(25(31)24(18-23)28(4,5)6)19-30-29(26(32)33,21-13-9-7-10-14-21)22-15-11-8-12-16-22;/h7-19,31H,1-6H3,(H,32,33);/q;+1/p-1/b30-19+;

Inchi Key

ZPKNCDGWTMRAKF-QUIZVCAPSA-M

Application

Hydroazidation Catalyst for Facile Preparation of Organoazides

SALDIPAC ligand used with Co(BF4)2 (cat. no. 399957) in a hydroazidation of unactivated olefins providing a simple synthesis of alkylazides.[1] The azides, in turn, can be used in a copper-catalyzed cycloaddition with acetylenes providing 1,4-disubstitutedtriazoles.[2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jérôme Waser et al.
Journal of the American Chemical Society, 127(23), 8294-8295 (2005-06-09)
Conversion of olefins to azides was achieved with high Markovnikov selectivity for a broad range of alkenes using 6 mol % Co(BF4).6H2O and ligand 1, with 3 equiv of TsN3 as nitrogen source and simple silanes (PhSiH3, TMDSO).
Synthesis, 3839-3839 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service