Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

676519

Sigma-Aldrich

6-(Piperidin-1-yl)pyridine-3-boronic acid pinacol ester

6-(Piperidin-1-yl)pyridine-3-boronic acid pinacol ester
1 of 1 reviewers received a sample product or took part in a promotion

97%

Synonym(s):

2-(Piperidin-1-yl)pyridine-5-boronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H25BN2O2
CAS Number:
Molecular Weight:
288.19
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

form

solid

mp

110-114 °C

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(nc2)N3CCCCC3

InChI

1S/C16H25BN2O2/c1-15(2)16(3,4)21-17(20-15)13-8-9-14(18-12-13)19-10-6-5-7-11-19/h8-9,12H,5-7,10-11H2,1-4H3

Inchi Key

MLJZIQHWAUYAPA-UHFFFAOYSA-N

Application

6-(Piperidin-1-yl)pyridine-3-boronic acid pinacol ester can be used as a reactant to synthesize:
  • Aromatic copolyamides applicable as fluorescent chemosensors for the detection of Cr, Fe, and Cu ions in organic-aqueous solutions.[1]
  • Purine based P2X7 receptor antagonists.[2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and SAR development of novel P2X7 receptor antagonists for the treatment of pain: Part 1
Matasi JJ, et al.
Bioorganic & medicinal chemistry letters, 21(12), 3805-3808 (2011)
Functional fluorescent aramids: aromatic polyamides containing a dipicolinic acid derivative as luminescent converters and sensory materials for the fluorescence detection and quantification of Cr (VI), Fe (III) and Cu (II)
Barrio-Manso J, et al.
Polym. Chem., 4(15), 4256-4264 (2013)

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. boston
    • Review 1
    • Votes 0
    5 out of 5 stars.

    6-(Piperidin-1-yl)pyridine-3-boronic acid pinacol ester

    This building block was of good quality and I would order it again.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service