Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

676470

Sigma-Aldrich

Nafion perfluorinated membrane

thickness 0.002 in.

Synonym(s):

NR-212, Perfluoro-3,6-dioxa-4-methyl-7-octenesulfonic polymer, Perfluoro[2-(2-sulfonylfluoroethoxy)propyl vinyl ether]-tetrafluoroethylene, Tetrafluoroethylene-perfluoro-3,6-dioxa-4-methyl-7-octenesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C7HF13O5S . C2F4)x
CAS Number:
MDL number:
UNSPSC Code:
26111700
eCl@ss:
32110203
NACRES:
NA.23

eq. wt.

1,100

L × W

12 in. × 12 in.

thickness

0.002 in.

solubility

H2O: insoluble

InChI

1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);

InChI key

FOYUGSIADQEOEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nafion NRE-212 membranes may be used in the investigation of oxygen reduction reaction (ORR) activity of Pt/C catalyst.[1]

Application

Carbon nanotube reinforced Nafion composites may be prepared to be used in H2O2 fuel cells.[2]
Nafion is a perfluorinated ion-exchange membrane which has a wide variety of commercial uses. The acidic (hydrogen ion) form and derivatives are solid, superacid catalysts useful in a wide variety of synthetic applications.
Nafion perfluorinated membrane is mainly used as a proton exchange membrane (PEM) for fuel cell applications. It has good proton conductivity and good physio-chemical stability. It can also be used as a reference membrane to compare and prepare high-performance composite membranes.[3][4][2]

Packaging

Darkening/browning is common for pieces stored in a general room environment for an extended period of time (several months), due to organics pick-up. Verify product is kept sealed during storage.
· Keep pieces sealed and protected from general exposure will reduce the effect. Recommend long term storage in air-tight bags.
· Discoloration does not affect performance and can be removed by soaking in dilute nitric acid.

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pt/SiO2 catalyst as an addition to Nafion/PTFE self-humidifying composite membrane
Wang L, et al.
Journal of Power Sources, 161(1), 61-67 (2006)
Novel multilayer Nafion/SPI/Nafion composite membrane for PEMFCs
Wang L, et al.
Journal of Power Sources, 164(1), 80-85 (2007)
Carbon nanotubes reinforced Nafion composite membrane for fuel cell applications.
Liu YH, et al.
Electrochemical and Solid-State Letters, 9(7), A356-A359 (2006)
Effect of methanol concentration on oxygen reduction reaction activity of Pt/C catalysts.
Shuihua, T. A. N. G., et al.
Chinese Journal of Catalysis, 34(6), 1105-1111 (2013)
Min-Ho Park et al.
Advanced materials (Deerfield Beach, Fla.), 27(29), 4308-4314 (2015-06-24)
A novel flexible encapsulation method (Flex Lami-capsulation) is reported, which can be applied in the roll-to-roll process for mass production of organic electronic devices. Flex Lami-capsulation is very simple, fast, and getter-free, and is as effective as glass encapsulation. Use

Articles

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service