Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

675113

Sigma-Aldrich

1-(1-Bromovinyl)-4-chlorobenzene

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6BrCl
CAS Number:
Molecular Weight:
217.49
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

90%

impurities

2-Boromo-1-(4-chlorophenyl)ethanone

refractive index

n20/D 1.605

density

1.539 g/mL at 25 °C

SMILES string

Clc1ccc(cc1)C(Br)=C

InChI

1S/C8H6BrCl/c1-6(9)7-2-4-8(10)5-3-7/h2-5H,1H2

InChI key

ZVDIEXFKTMIHIU-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service