Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

674214

Sigma-Aldrich

1-Hydroxy-3-(isothiocyanato)-1,1,3,3-tetrabutyldistannoxane dimer

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H37NO2SSn2)2
CAS Number:
Molecular Weight:
1113.93
UNSPSC Code:
12352300
PubChem Substance ID:

assay

97%

form

solid

reaction suitability

core: tin
reagent type: catalyst

mp

136-151 °C

SMILES string

CCCC[Sn](O)(CCCC)O[Sn](CCCC)(CCCC)N=C=S

InChI

1S/4C4H9.CNS.H2O.O.2Sn/c4*1-3-4-2;2-1-3;;;;/h4*1,3-4H2,2H3;;1H2;;;/q;;;;-1;;;2*+1/p-1

InChI key

VGXGQBKDGBKKRA-UHFFFAOYSA-M

Application

Catalyst for esterification

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Otera, J.; Yano, T.; Okawara, R.
Chemistry Letters (Jpn), 14, 901-901 (1985)
Otera, J.; Danoh, N.; Nozaki, H.
The Journal of Organic Chemistry, 56, 5307-5307 (1991)
Otera, J.
Chemical Reviews, 93, 1449-1449 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service